Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein phosphatase methylesterase 1 | PPME1 | PME-1 | Anti-sense | Chr. 11 - 74170891 - 74254703 on NCBI Build GRCh38 | |
Downstream | prolyl 4-hydroxylase subunit alpha 3 | P4HA3 | 62.9kb | Sense | Chr. 11 - 74259923 - 74311676 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |