Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010171.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010171.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010171.hg.1 |
TC10000840.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161601.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161601.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161601.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161601.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161601.hg.1 |
TC1000009000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000333254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000463594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
ENST00000497093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009000.hg.1 |
NM_198515.2 |
Refseq |