Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000422.hg.1 |
TC03000017.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000422.hg.1 |
ENST00000256497.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000422.hg.1 |
ENST00000419550.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000422.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000422.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144606.hg.1 |
TC0300006491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000256497.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000434243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000443790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000445686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000465369.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
ENST00000492751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006491.hg.1 |
NM_014674.2 |
Refseq |