Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003467.2 | NP_003458.1 | 1 - 2 | 112 | 79 | |
GenBank mRNA | AF348491.1 | - | 1 - 2 | 60 | 79 | |
AK129916.1 | - | 1 - 2 | 106 | 79 | ||
AK296674.1 | - | 1 - 2 | 106 | 79 | ||
AK312244.1 | - | 1 - 2 | 106 | 79 | ||
BC020968.2 | - | 1 - 2 | 70 | 3846345 | 79 | |
D10924.1 | - | 1 - 2 | 93 | 79 | ||
L06797.1 | - | 1 - 2 | 89 | 79 | ||
M99293.1 | - | 1 - 2 | 101 | 79 | ||
X71635.1 | - | 1 - 2 | 84 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |