Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011163.hg.1 |
ENST00000385191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011163.hg.1 |
NR_029677.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
TC03000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
ENST00000354773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
ENST00000411757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007135.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NM_145262.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026700.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
NR_026702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153730.hg.1 |
TC0300007472.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011163.hg.1 |
ENST00000385191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011163.hg.1 |
NR_029677.1 |
Refseq |