Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017613.hg.1 |
TC01001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017613.hg.1 |
ENST00000234739.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017613.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017614.hg.1 |
TC01001113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017614.hg.1 |
ENST00000234739.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017614.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017614.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000497938.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009788.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000234739.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000473292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
ENST00000622552.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
NM_004326.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100170561.hg.1 |
TC0100009788.hg.1 |
Clariom D TC |