Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
TC01002153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
ENST00000348333.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
ENST00000356876.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
ENST00000377782.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
ENST00000473343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
NM_003790.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
NM_148965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033177.hg.1 |
NM_148967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000348333.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000356876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000377782.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000414040.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000453260.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000469691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000473343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000480393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000481401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000485036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000513135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_003790.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_148965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_148967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000348333.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000356876.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000377782.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000414040.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000453260.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000469691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000473343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000480393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000481401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000485036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
ENST00000513135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_003790.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_148965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
NM_148967.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241320.hg.1 |
TC0100018398.hg.1 |
Clariom D TC |