Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
ENST00000332811.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
ENST00000402174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
ENST00000462485.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
ENST00000544604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003471.hg.1 |
TC22000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003466.hg.1 |
ENST00000402174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003466.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003466.hg.1 |
TC22000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
ENST00000332811.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
ENST00000402174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
ENST00000462485.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
ENST00000544604.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003472.hg.1 |
TC22000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000418292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000418292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000585003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009245.hg.1 |
ENST00000585003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000418292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000418292.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000422972.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000451486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000458080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000585003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009245.hg.1 |
ENST00000585003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
TC2200009246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168373.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168370.hg.1 |
TC2200009246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168370.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168370.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168370.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168370.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
TC2200009246.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168374.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000402174.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000406323.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
ENST00000544604.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_001206998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009246.hg.1 |
NM_032173.3 |
Refseq |