Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005701.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005701.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005701.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005701.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005701.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005699.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
ENST00000545501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005702.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005696.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005696.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005696.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005697.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005697.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005697.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005697.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005703.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005703.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005703.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005703.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005703.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
TC02000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
ENST00000377990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
ENST00000545501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
ENST00000546106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005704.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154005.hg.1 |
TC0200007825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154008.hg.1 |
TC0200007825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000497039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
ENST00000537589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007825.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
ENST00000475851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154003.hg.1 |
TC0200007825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
ENST00000260569.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
ENST00000377990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
NM_015147.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154007.hg.1 |
TC0200007825.hg.1 |
Clariom D TC |