Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TONSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013432.4 | NP_038460.4 | 24 - 25 | 3822 | 96 | |
XM_011517048.2 | XP_011515350.1 | 17 - 18 | 2921 | 96 | ||
XM_011517049.2 | XP_011515351.1 | 17 - 18 | 2882 | 96 | ||
GenBank mRNA | AB209126.1 | - | 15 - 16 | 5909 | 96 | |
AK097406.1 | - | 3 - 4 | 1837 | 96 | ||
BC008782.2 | - | 23 - 24 | 3794 | 3628374 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |