Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012423.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
ENST00000375329.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195396.hg.1 |
TC1100012423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000236850.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000359492.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000375320.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
ENST00000375323.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
NM_000039.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195395.hg.1 |
TC1100012423.hg.1 |
Clariom D TC |