Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023591.hg.1 |
TC06001926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023591.hg.1 |
ENST00000369582.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023591.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023591.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000627552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012495.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000369582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000610310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000625577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000627148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
ENST00000630630.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
NM_000735.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
NM_001252383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186130.hg.1 |
TC0600012495.hg.1 |
Clariom D TC |