Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003034.hg.1 |
ENST00000377589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003034.hg.1 |
ENST00000377595.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003034.hg.1 |
ENST00000471792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003034.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003034.hg.1 |
TC13000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000606347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000607339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009979.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000377595.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000419068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000471792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
ENST00000618773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
NM_001270952.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
NM_006002.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166715.hg.1 |
TC1300009979.hg.1 |
Clariom D TC |