Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
ENST00000254963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
ENST00000542646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000658.hg.1 |
TC20000039.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
TC2000006558.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145068.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000254963.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
ENST00000399701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_001197327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006558.hg.1 |
NM_052970.4 |
Refseq |