Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011835.hg.1 |
ENST00000434758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011835.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011835.hg.1 |
TC11000951.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011836.hg.1 |
ENST00000434758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011836.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11011836.hg.1 |
TC11000951.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000526781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000529204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
ENST00000534360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_001195005.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008900.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165647.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165647.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165647.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165647.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165647.hg.1 |
TC1100008900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165644.hg.1 |
TC1100008900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
ENST00000434758.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
ENST00000530659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
NM_032930.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100165645.hg.1 |
TC1100008900.hg.1 |
Clariom D TC |