Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005708.hg.1 |
TC05000400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005708.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005708.hg.1 |
ENST00000535995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005708.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
ENST00000512531.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005709.hg.1 |
TC05000400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005709.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005709.hg.1 |
ENST00000535995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005709.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512258.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
ENST00000512531.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007919.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154259.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154259.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154259.hg.1 |
ENST00000265081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500154259.hg.1 |
NM_002439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500154259.hg.1 |
TC0500007919.hg.1 |
Clariom D TC |