Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013388.hg.1 |
ENST00000264819.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013388.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013388.hg.1 |
TC19000982.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000587966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000587966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172044.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172044.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172044.hg.1 |
TC1900009078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172044.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172044.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000264819.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000586994.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000587966.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000587966.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000592722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000619835.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
ENST00000621180.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009078.hg.1 |
NM_017550.2 |
Refseq |