Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024840.hg.1 |
ENST00000344935.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024840.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024840.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024840.hg.1 |
TC17001848.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024841.hg.1 |
ENST00000344935.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024841.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024841.hg.1 |
TC17001848.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700194508.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194508.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194508.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194508.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194508.hg.1 |
TC1700011656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194506.hg.1 |
TC1700011656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700194507.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194507.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194507.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700194507.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700194507.hg.1 |
TC1700011656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000344935.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
ENST00000397671.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011656.hg.1 |
NM_001129885.1 |
Refseq |