Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000357716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389335.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000389336.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000402647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000435689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000440923.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000443259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000445087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000445087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000445087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000445087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000445087.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000448328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000448328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000448328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000448328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000448328.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000458641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000461677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000461677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000461677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000461677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000461677.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000463676.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000467425.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000471242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000471242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
ENST00000486284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_000844.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006513.hg.1 |
NM_181874.2 |
Refseq |