Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153410.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153410.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153410.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153410.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153410.hg.1 |
TC0500007811.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000504514.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000506596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000594319.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
ENST00000601380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007811.hg.1 |
NM_001276713.1 |
Refseq |