Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
TC08001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
ENST00000265572.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
ENST00000396798.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
ENST00000520287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
ENST00000524278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015670.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
TC08001212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
ENST00000265572.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
ENST00000396798.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
ENST00000520287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
ENST00000524278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08015671.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800170904.hg.1 |
TC0800010441.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000265572.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000520287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000522508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000524278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000612786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
ENST00000613482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_000912.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010441.hg.1 |
NM_001282904.1 |
Refseq |