Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014786.3 | NP_055601.2 | 12 - 13 | 4516 | 74 | |
XM_017018623.1 | XP_016874112.1 | 12 - 13 | 2907 | 74 | ||
XM_017018624.1 | XP_016874113.1 | 11 - 12 | 1986 | 74 | ||
GenBank mRNA | AB002335.1 | - | 12 - 13 | 3290 | 74 | |
AF378754.1 | - | 12 - 13 | 4510 | 74 | ||
BC137228.1 | - | 12 - 13 | 4516 | 9021225 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |