Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201465.1 | NP_001188394.1 | 2 - 3 | 371 | 112 | |
NM_138966.3 | NP_620416.1 | 2 - 3 | 371 | 112 | ||
XM_005266774.4 | XP_005266831.1 | 2 - 3 | 309 | 112 | ||
XM_017026017.1 | XP_016881506.1 | 2 - 3 | 233 | 112 | ||
XM_017026018.1 | XP_016881507.1 | 2 - 3 | 233 | 112 | ||
XM_017026022.1 | XP_016881511.1 | 2 - 3 | 151 | 112 | ||
XM_017026023.1 | XP_016881512.1 | 2 - 3 | 151 | 112 | ||
XM_017026024.1 | XP_016881513.1 | 2 - 3 | 366 | 112 | ||
GenBank mRNA | AF448838.1 | - | 2 - 3 | 377 | 112 | |
BC050329.1 | - | 2 - 3 | 373 | 4821317 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |