Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003356.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003356.hg.1 |
ENST00000549706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003356.hg.1 |
ENST00000553030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003356.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003356.hg.1 |
TC19000227.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
TC1900007090.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150957.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000042931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000549706.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000552539.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
ENST00000553030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007090.hg.1 |
NM_017682.2 |
Refseq |