Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 207 | TMEM207 | UNQ846 | -149.9kb | Sense | Chr. 3 - 190428655 - 190449876 on NCBI Build GRCh38 |
Downstream | geminin coiled-coil domain containing | GMNC | GEMC1 | 241.5kb | Sense | Chr. 3 - 190843271 - 190864200 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |