Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017816.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017816.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017816.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017816.hg.1 |
TC15001836.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017817.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017817.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017817.hg.1 |
TC15001836.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017815.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017815.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017815.hg.1 |
TC15001836.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178392.hg.1 |
TC1500010404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178391.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178391.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178391.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178391.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178391.hg.1 |
TC1500010404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500178394.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178394.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178394.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178394.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178394.hg.1 |
TC1500010404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
ENST00000559894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010404.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178393.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178393.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178393.hg.1 |
ENST00000300057.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500178393.hg.1 |
NM_018670.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500178393.hg.1 |
TC1500010404.hg.1 |
Clariom D TC |