Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260512.1 | NP_001247441.1 | 1 - 2 | 63 | 75 | |
NM_001267598.1 | NP_001254527.1 | 1 - 2 | 63 | 75 | ||
NM_020300.4 | NP_064696.1 | 1 - 2 | 63 | 75 | ||
NR_048545.1 | 1 - 2 | 63 | 75 | |||
XM_006719079.1 | XP_006719142.1 | 1 - 2 | 54 | 75 | ||
XM_011520673.1 | XP_011518975.1 | 1 - 2 | 54 | 75 | ||
XM_011520674.2 | XP_011518976.1 | 1 - 2 | 54 | 75 | ||
GenBank mRNA | AK291148.1 | - | 1 - 2 | 52 | 75 | |
BC005923.1 | - | 1 - 2 | 31 | 4297300 | 75 | |
BG617746.1 | - | 1 - 2 | 64 | 75 | ||
BG778603.1 | - | 1 - 2 | 64 | 75 | ||
H49331.1 | - | 1 - 2 | 61 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |