Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004019.1 | NP_001004019.1 | 9 - 10 | 2419 | 67 | |
NM_001165035.1 | NP_001158507.1 | 9 - 10 | 2676 | 67 | ||
XM_006713026.2 | XP_006713089.1 | 8 - 9 | 2328 | 67 | ||
GenBank mRNA | AK300913.1 | - | 4 - 5 | 429 | 67 | |
AK304827.1 | - | 9 - 10 | 2676 | 67 | ||
AL050095.1 | - | 3 - 4 | 330 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |