Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000263150.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000358220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000381329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000436154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000482165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
ENST00000620998.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006466.hg.1 |
NM_014023.3 |
Refseq |