Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161664.hg.1 |
TC0300008686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000290913.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000503119.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000504973.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000508789.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000510044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000513253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000514908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
ENST00000515867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008686.hg.1 |
NM_032343.2 |
Refseq |