Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025668.hg.1 |
TC02001600.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025668.hg.1 |
ENST00000355549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025668.hg.1 |
ENST00000381323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025668.hg.1 |
ENST00000406434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025668.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200185939.hg.1 |
TC0200011837.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000381323.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000406434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000445605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000451689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
ENST00000469507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011837.hg.1 |
NM_030797.3 |
Refseq |