Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009670.hg.1 |
TC0X000745.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009670.hg.1 |
ENST00000370264.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009670.hg.1 |
ENST00000370265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009670.hg.1 |
ENST00000447306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009670.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009666.hg.1 |
TC0X000745.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009666.hg.1 |
ENST00000370265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009666.hg.1 |
ENST00000447306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009666.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009665.hg.1 |
TC0X000745.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009665.hg.1 |
ENST00000370265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009665.hg.1 |
ENST00000447306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009665.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009667.hg.1 |
TC0X000745.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009667.hg.1 |
ENST00000370265.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009667.hg.1 |
ENST00000447306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009667.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159658.hg.1 |
TC0X00008764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159661.hg.1 |
TC0X00008764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000593810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008764.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159657.hg.1 |
TC0X00008764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
ENST00000424805.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
ENST00000619635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
NM_001282535.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
NM_013364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159659.hg.1 |
TC0X00008764.hg.1 |
Clariom D TC |