Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006612.hg.1 |
ENST00000361677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006612.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006612.hg.1 |
TC17000543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006614.hg.1 |
ENST00000361677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006614.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006614.hg.1 |
TC17000543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009121.hg.1 |
ENST00000603595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009121.hg.1 |
ENST00000603595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000590836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
ENST00000590836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157926.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157926.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157926.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157926.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157926.hg.1 |
TC1700007929.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009121.hg.1 |
ENST00000603595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009121.hg.1 |
ENST00000603595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000590836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
ENST00000590836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007929.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
ENST00000589705.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157925.hg.1 |
TC1700007929.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157927.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157927.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157927.hg.1 |
ENST00000361677.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157927.hg.1 |
NM_173079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157927.hg.1 |
TC1700007929.hg.1 |
Clariom D TC |