Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007080.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007080.hg.1 |
ENST00000455537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007080.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007080.hg.1 |
TC12000534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
ENST00000552227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012651.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200728.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200728.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200728.hg.1 |
ENST00000286452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200200728.hg.1 |
NM_004984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200200728.hg.1 |
TC1200012651.hg.1 |
Clariom D TC |