Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
TC09000677.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
ENST00000277480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
ENST00000373013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
ENST00000373017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
ENST00000540948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008149.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
TC09000677.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000277480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000373013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000373017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000470902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
ENST00000540948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008148.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
TC09000677.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
ENST00000277480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
ENST00000373013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
ENST00000373017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
ENST00000470902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
ENST00000540948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008147.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159248.hg.1 |
TC0900008847.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159246.hg.1 |
TC0900008847.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159249.hg.1 |
TC0900008847.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000277480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000372998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000373017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000470902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000487719.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000488391.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
ENST00000494317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008847.hg.1 |
NM_005564.4 |
Refseq |