Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023726.hg.1 |
TC07001570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023726.hg.1 |
ENST00000284136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023726.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023726.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023727.hg.1 |
TC07001570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023727.hg.1 |
ENST00000284136.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023727.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023727.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183345.hg.1 |
TC0700011675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183346.hg.1 |
TC0700011675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000284136.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000444867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000463315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
ENST00000484038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011675.hg.1 |
NM_152754.2 |
Refseq |