Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025080.hg.1 |
TC01001624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025080.hg.1 |
ENST00000235382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025080.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025079.hg.1 |
TC01001624.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025079.hg.1 |
ENST00000235382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025079.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180850.hg.1 |
TC0100011022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180851.hg.1 |
TC0100011022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000235382.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000464302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000483295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
ENST00000487236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011022.hg.1 |
NM_002923.3 |
Refseq |