Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002152.2 | NP_002143.1 | 3 - 4 | 2156 | 78 | |
GenBank mRNA | BC069795.1 | - | 3 - 4 | 2139 | 7262494 | 78 |
BC069802.1 | - | 3 - 4 | 2139 | 7262506 | 78 | |
BC094691.1 | - | 3 - 4 | 2156 | 30529366 | 78 | |
BC112355.1 | - | 3 - 4 | 2121 | 40034092 | 78 | |
M60052.1 | - | 3 - 4 | 2139 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |