Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017661.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233327.hg.1 |
TC0100017661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233328.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233328.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233328.hg.1 |
TC0100017661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233324.hg.1 |
TC0100017661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000284617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000366687.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
ENST00000483092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
NM_145257.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233323.hg.1 |
TC0100017661.hg.1 |
Clariom D TC |