Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001782.hg.1 |
TC03000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001782.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001782.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013793.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
ENST00000306077.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
ENST00000432444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
NM_024334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300203496.hg.1 |
TC0300013793.hg.1 |
Clariom D TC |