Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077351.1 | NP_001070819.1 | 4 - 5 | 425 | 66 | |
XM_011536890.2 | XP_011535192.1 | 5 - 6 | 508 | 66 | ||
XM_011536894.2 | XP_011535196.1 | 5 - 6 | 508 | 66 | ||
XM_011536900.1 | XP_011535202.1 | 4 - 5 | 422 | 66 | ||
XM_011536902.1 | XP_011535204.1 | 3 - 4 | 274 | 66 | ||
XM_011536904.1 | XP_011535206.1 | 3 - 4 | 261 | 66 | ||
XM_017021401.1 | XP_016876890.1 | 4 - 5 | 422 | 66 | ||
XM_017021402.1 | XP_016876891.1 | 4 - 5 | 333 | 66 | ||
XM_017021405.1 | XP_016876894.1 | 4 - 5 | 422 | 66 | ||
XM_017021406.1 | XP_016876895.1 | 4 - 5 | 840 | 66 | ||
XM_017021407.1 | XP_016876896.1 | 4 - 5 | 349 | 66 | ||
XM_017021409.1 | XP_016876898.1 | 4 - 5 | 840 | 66 | ||
XM_017021413.1 | XP_016876902.1 | 3 - 4 | 236 | 66 | ||
XM_017021414.1 | XP_016876903.1 | 3 - 4 | 274 | 66 | ||
XM_017021417.1 | XP_016876906.1 | 3 - 4 | 261 | 66 | ||
GenBank mRNA | AF275678.1 | - | 4 - 5 | 379 | 66 | |
AK125593.1 | - | 4 - 5 | 387 | 66 | ||
AK225792.1 | - | 4 - 5 | 387 | 66 | ||
AL834198.1 | - | 4 - 5 | 386 | 66 | ||
BC106012.1 | - | 2 - 3 | 178 | 66 | ||
BX161440.1 | - | 4 - 5 | 358 | 66 | ||
KU178688.1 | - | 3 - 4 | 226 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |