Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002830.3 | NP_002821.1 | 18 - 19 | 2429 | 66 | |
XM_011511560.2 | XP_011509862.1 | 18 - 19 | 2334 | 66 | ||
XM_011511562.2 | XP_011509864.1 | 17 - 18 | 2268 | 66 | ||
XM_011511563.2 | XP_011509865.1 | 8 - 9 | 1792 | 66 | ||
XM_017004596.1 | XP_016860085.1 | 18 - 19 | 2355 | 66 | ||
XM_017004597.1 | XP_016860086.1 | 18 - 19 | 1861 | 66 | ||
XM_017004598.1 | XP_016860087.1 | 18 - 19 | 2334 | 66 | ||
XM_017004599.1 | XP_016860088.1 | 18 - 19 | 1840 | 66 | ||
XM_017004600.1 | XP_016860089.1 | 18 - 19 | 1819 | 66 | ||
XM_017004601.1 | XP_016860090.1 | 17 - 18 | 2310 | 66 | ||
XM_017004602.1 | XP_016860091.1 | 17 - 18 | 2289 | 66 | ||
XM_017004603.1 | XP_016860092.1 | 17 - 18 | 1774 | 66 | ||
XM_017004604.1 | XP_016860093.1 | 17 - 18 | 2244 | 66 | ||
XM_017004605.1 | XP_016860094.1 | 17 - 18 | 2140 | 66 | ||
XM_017004606.1 | XP_016860095.1 | 17 - 18 | 2265 | 66 | ||
XM_017004607.1 | XP_016860096.1 | 15 - 16 | 2026 | 66 | ||
XM_017004608.1 | XP_016860097.1 | 15 - 16 | 1983 | 66 | ||
GenBank mRNA | AK301218.1 | - | 6 - 7 | 645 | 66 | |
AK314836.1 | - | 18 - 19 | 2241 | 66 | ||
BC010674.1 | - | 18 - 19 | 2085 | 3853914 | 66 | |
BX648614.1 | - | 12 - 13 | 2663 | 66 | ||
M68941.1 | - | 18 - 19 | 2429 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons |