Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
TC02002429.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
ENST00000295087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
ENST00000428992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
ENST00000452215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02038313.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006902.hg.1 |
ENST00000430731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006902.hg.1 |
ENST00000430731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200227870.hg.1 |
TC0200016740.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2200006902.hg.1 |
ENST00000430731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006902.hg.1 |
ENST00000430731.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000295087.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000428992.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000458140.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
ENST00000487723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_001037174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_012097.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016740.hg.1 |
NM_177985.2 |
Refseq |