Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500168541.hg.1 |
TC0500009725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000261951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000393356.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000502447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000504343.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000507016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000510148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
ENST00000618123.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009725.hg.1 |
NM_001303241.1 |
Refseq |