Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023331.hg.1 |
TC03001362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023331.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023331.hg.1 |
ENST00000395869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023331.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179988.hg.1 |
TC0300010962.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000292327.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000395869.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
ENST00000431168.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010962.hg.1 |
NM_000258.2 |
Refseq |