Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006496.3 | NP_006487.1 | 5 - 6 | 739 | 80 | |
GenBank mRNA | AF493907.1 | - | 5 - 6 | 582 | 80 | |
AK311691.1 | - | 6 - 7 | 602 | 80 | ||
AK312252.1 | - | 5 - 6 | 668 | 80 | ||
BC025285.1 | - | 5 - 6 | 692 | 5433334 | 80 | |
BT019973.1 | - | 5 - 6 | 582 | 80 | ||
BT019974.1 | - | 5 - 6 | 582 | 80 | ||
J03005.1 | - | 5 - 6 | 590 | 80 | ||
J03198.1 | - | 5 - 6 | 644 | 80 | ||
J03238.1 | - | 5 - 6 | 672 | 80 | ||
M27543.1 | - | 5 - 6 | 624 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |