Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000354656.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000355684.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000356109.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
ENST00000483826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_001164276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011880.hg.1 |
NM_016264.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204489.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204489.hg.1 |
TC1900011880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900204489.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204490.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204490.hg.1 |
ENST00000397742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204490.hg.1 |
TC1900011880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900204490.hg.1 |
ENST00000393337.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204490.hg.1 |
ENST00000397742.7 |
Ensembl |