Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01022239_m1
See other RBFOX2 GE Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01022239_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005261428.2
XP_005261485.1
10 - 11
1570
94
XM_005261429.2
XP_005261486.1
10 - 11
1567
94
XM_005261435.1
XP_005261492.1
9 - 10
1271
94
XM_006724185.2
XP_006724248.1
10 - 11
1582
94
XM_006724186.2
XP_006724249.1
10 - 11
1579
94
XM_006724189.3
XP_006724252.1
10 - 11
1275
94
XM_006724190.1
XP_006724253.1
10 - 11
969
94
XM_006724191.1
XP_006724254.1
9 - 10
1274
94
XM_006724192.2
XP_006724255.1
9 - 10
1104
94
XM_006724194.2
XP_006724257.1
9 - 10
1046
94
XM_017028686.1
XP_016884175.1
10 - 11
961
94
XM_017028687.1
XP_016884176.1
10 - 11
1142
94
XM_017028689.1
XP_016884178.1
10 - 11
1035
94
XM_017028690.1
XP_016884179.1
9 - 10
1024
94
XM_017028691.1
XP_016884180.1
9 - 10
1021
94
XM_017028693.1
XP_016884182.1
9 - 10
1262
94
XM_017028694.1
XP_016884183.1
9 - 10
1259
94
XM_017028697.1
XP_016884186.1
10 - 11
2051
94
GenBank mRNA
AB649123.1
-
9 - 10
902
94
AM419009.1
-
9 - 10
914
94
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Amplicon length between 86 and 100
Homo sapiens
Probe spans exons
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0