Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021037.hg.1 |
ENST00000263847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021037.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021037.hg.1 |
TC11001827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010976.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182781.hg.1 |
TC1100010976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000263847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000525357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
ENST00000528903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
NM_002556.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100182780.hg.1 |
TC1100010976.hg.1 |
Clariom D TC |