Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK301027.1 | - | 4 - 5 | 536 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |